Learn More
Research Products International Corp Dish and Cover for 20 Slide Staining Rack
SDP
Catalog No. 501978103
Encompass
This 20-slide unit is the standard for manual staining procedures. The removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carryover. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back of standard size:
3 x 1 Inch (75 x 25 mm)
3 x 1 1/2 Inch (75 x 38 mm)
3 x 2 Inch (75 x 51 mm)
Catalog No. 50-197-8103 Supplier Research Products International Corp Supplier No. 144203
This 20-slide unit is the standard for manual staining procedures. The removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carryover. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back of standard size:
3 x 1 Inch (75 x 25 mm)
3 x 1 1/2 Inch (75 x 38 mm)
3 x 2 Inch (75 x 51 mm)