Learn More
MSC Vikan Dish Brush, Scraping Edge, 11 inch, Medium Bristle, Yellow
Catalog No. 502835627
Encompass
This dish brush is designed for detail cleaning of machinery and equipment. It features a scraping edge, 11 inch length, medium bristles, and is yellow in color.
  • Purpose-built for food manufacturing, food retail, restaurants, and food service where hygiene and food safety are critical.
  • Medium bristles work well scrubbing with a cleaning solution.
  • Ergonomic design enhances comfort and reduces worker strain.
  • Curved brush head and bristles clean inside curved glassware and bowls.
  • Scraping edge loosens stuck-on soils.
  • Durable construction provides long-lasting performance with daily use.
  • Easy to clean and maintain for hygiene control.
  • Color-coded for use with hygienic zoning plans and 5S lean programs.
Catalog No. 50-283-5627 Supplier MSC Supplier No. 47175401
This dish brush is designed for detail cleaning of machinery and equipment. It features a scraping edge, 11 inch length, medium bristles, and is yellow in color.
  • Purpose-built for food manufacturing, food retail, restaurants, and food service where hygiene and food safety are critical.
  • Medium bristles work well scrubbing with a cleaning solution.
  • Ergonomic design enhances comfort and reduces worker strain.
  • Curved brush head and bristles clean inside curved glassware and bowls.
  • Scraping edge loosens stuck-on soils.
  • Durable construction provides long-lasting performance with daily use.
  • Easy to clean and maintain for hygiene control.
  • Color-coded for use with hygienic zoning plans and 5S lean programs.