Learn More
Hach Company Dish, Evaporating, Porcelain, 70 mL
Catalog No. 501994361 Shop All Hach Company Products
Encompass
Deep-form, round bottom with lip. Glazed except for rim. Temperatures to 1500°C (max change = ±200°C/hr).
Catalog No. 50-199-4361 Supplier Hach Company Supplier No. 52590
Deep-form, round bottom with lip. Glazed except for rim. Temperatures to 1500°C (max change = ±200°C/hr).