Learn More
Chemglass Life Sciences Glass-bottom culture dish, clear, 35 x 10 mm, TC-treated
SDP
Catalog No. 502733284
Encompass_Preferred
Glass-bottom culture dish for live cell observation and microscopy. The optical-grade glass bottom and TC-treated surface enable high-resolution imaging and cell attachment, while e-beam sterilization and medical-grade adhesive ensure non-cytotoxic, ready-to-use components.
  • Optical-grade glass bottom, 35 x 10 mm, 15 mm well diameter.
  • Optical glass thickness 0.16-0.19 mm.
  • Tissue-culture treated surface to promote cell attachment.
  • E-beam sterilized for aseptic use.
  • Medical-grade adhesive to ensure non-cytotoxic bonding.
  • Pack quantity: 12.
Catalog No. 50-273-3284 Supplier Chemglass Life Sciences Supplier No. CLS1811001
Glass-bottom culture dish for live cell observation and microscopy. The optical-grade glass bottom and TC-treated surface enable high-resolution imaging and cell attachment, while e-beam sterilization and medical-grade adhesive ensure non-cytotoxic, ready-to-use components.
  • Optical-grade glass bottom, 35 x 10 mm, 15 mm well diameter.
  • Optical glass thickness 0.16-0.19 mm.
  • Tissue-culture treated surface to promote cell attachment.
  • E-beam sterilized for aseptic use.
  • Medical-grade adhesive to ensure non-cytotoxic bonding.
  • Pack quantity: 12.