Learn More
Chemglass Life Sciences Chemglass Glass-Bottom Culture Dish, 35 x 10 mm, 20 mm Glass, TC-Treated
SDP
Catalog No. 502733283
Encompass_Preferred
Glass-bottom culture dish designed for live-cell microscopy and imaging. The dish has a 35 x 10 mm footprint with a 20 mm optical-grade glass bottom (0.16-0.19 mm thickness), medical-grade adhesive for non-cytotoxic bonding, and E-beam sterilization.
  • For confocal, differential interference contrast, polarized light, and phase contrast microscopy.
  • Optical-grade glass bottom, 0.16-0.19 mm thick.
  • 20 mm clear observation area.
  • Medical-grade adhesive ensures non-cytotoxic bonding.
  • Sterilized by E-beam.
  • Packaged in packs of 10.
Catalog No. 50-273-3283 Supplier Chemglass Life Sciences Supplier No. CLS1811002
Glass-bottom culture dish designed for live-cell microscopy and imaging. The dish has a 35 x 10 mm footprint with a 20 mm optical-grade glass bottom (0.16-0.19 mm thickness), medical-grade adhesive for non-cytotoxic bonding, and E-beam sterilization.
  • For confocal, differential interference contrast, polarized light, and phase contrast microscopy.
  • Optical-grade glass bottom, 0.16-0.19 mm thick.
  • 20 mm clear observation area.
  • Medical-grade adhesive ensures non-cytotoxic bonding.
  • Sterilized by E-beam.
  • Packaged in packs of 10.