Learn More
Hach Company Dish, Petri, 9 x 50 mm, 100/pack
Supplier:  Hach Company 1485299
Encompass
Suitable for 47 mm membrane filters, these 9 x 50-mm polystyrene dishes are presterilized and disposable. Double rim ensures tight seal. An area of the lid is frosted for identification.
Catalog No. 50-199-5135