Learn More
Electron Microscopy Sciences Ilford Dish/Tray Developer for Multigrade Paper
SDP
Supplier:  Electron Microscopy Sciences 8305005
Encompass
MULTIGRADE is a liquid developer which is concentrated and it is suitable for all black and white papers. It can be used at a standard dilution of 1+9 or at 1+14 for greater development control and economy. MULTIGRADE developer is clean working and has excellent keeping properties and gives a neutral image tone with most papers. MULTIGRADE is designed for use at ambient room temperatures, normally 20 degree C/68 degree F. We do not recommend its use for high temperature or machine processing applications. It is not suitable for film developing.
Catalog No. 50-313-74