Learn More
Thomas Scientific Weigh Boat, black, 20ml, 45 x 45mm, square
SDP
Catalog No. 501458331
Encompass_Preferred
MTC Bio Weigh Boats are made of antistatic polystyrene and will withstand temperatures to 90ÝC, as well as exposure to diluted acids, alcohols, bases and aqueous solutions. Their flexibility and smooth interior surfaces with rounded corners allow for easy decanting of powdered samples. Their flat bottoms ensure stability and prevent tipping. Weigh Boats are available in the traditional square shape as well as in a new diamond shape for easier pouring. Originally designed as weighing containers, they have proven ideal for dispensing & storage and as disposable mixing/quick freezing trays. They are available in black or white for improved sample contrast/visibility and packaged in ultra-compact dispenser packs. Available in diamond or square shape.
Catalog No. 50-145-8331 Supplier Thomas Scientific Supplier No. 1158Q72
MTC Bio Weigh Boats are made of antistatic polystyrene and will withstand temperatures to 90ÝC, as well as exposure to diluted acids, alcohols, bases and aqueous solutions. Their flexibility and smooth interior surfaces with rounded corners allow for easy decanting of powdered samples. Their flat bottoms ensure stability and prevent tipping. Weigh Boats are available in the traditional square shape as well as in a new diamond shape for easier pouring. Originally designed as weighing containers, they have proven ideal for dispensing & storage and as disposable mixing/quick freezing trays. They are available in black or white for improved sample contrast/visibility and packaged in ultra-compact dispenser packs. Available in diamond or square shape.