Learn More
Enzo Life Sciences Dishevelled-1 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences BMLDA41700100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 557-571 (CS557RRAPGREKERRAAG571) of human dishevelled 1 protein. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat. Applications: WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 10mM sodium azide.
Catalog No. 50-201-0502