Learn More
Enzo Life Sciences Dishevelled-2 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences BMLDA42700100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 600-614 (CG600AGRTGRPEERAPES614) of human and mouse dishevelled 2 protein. The amino acid sequence does not exist in human or mouse dvl-1 or dvl-3. Source: Purified from rabbit serum. Species reactivity: Human, Mouse. Applications: IHC, WB. Formulation: Liquid. In PBS, pH 7.4, containing 10mM sodium azide.
Catalog No. 50-201-0504