Learn More
Cayman Chemical DIsIalognglIosIde MIxture 1mL
Supplier: Cayman Chemical 293601 ML
A mixture of the disialogangliosides GD1a, GD1b, and GD3; intended for use as a reference standard for the detection of disialogangliosides in a variety of sample types by TLC, LC-, or GC-MS