Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Disilane

Catalog No. 46304320G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-304-320G 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46-304-320G Supplier Sigma Aldrich Supplier No. 46304320G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Disilane

Specifications

CAS 1590-87-0
Melting Point -132.6°C
Boiling Point °C to 14.5°C
Molecular Formula H6Si2
Linear Formula Si2H6
MDL Number MFCD00054678
Quantity 20 g
Synonym Disilicane; Disilicoethane; Disilicon hexahydride; Silicoethane; Silicon hydride (Si2 H6)
Molecular Weight (g/mol) 62.22
Percent Purity ≥99.998%chemical purity basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.