Learn More
PENTAIR AQUATIC ECO SYSTEMS DISINFECTANT 10LB VIRKON AQUAT
Catalog No. NC2259246
Encompass
Disinfectant 10LB Virkon AquaticThis is a highly effective disinfectant specifically formulated for use in aquaculture facilities. Generally used for cleaning equipment and instruments, but may also be used in footbaths. Ideal for controlling a wide variety of viral, bacterial and fungal pathogens.Mix at a rate of 1.3 ounces per gallon of water. One gallon of solution treats 135 sq.ft.; 10-lb container makes 123 gallons of solution. Solution remains stable for seven days. EPA-registered (registration pending in California).
This item is not returnable. View return policy
Catalog No. NC2259246 Supplier PENTAIR AQUATIC ECO SYSTEMS Supplier No. VKS10
Disinfectant 10LB Virkon AquaticThis is a highly effective disinfectant specifically formulated for use in aquaculture facilities. Generally used for cleaning equipment and instruments, but may also be used in footbaths. Ideal for controlling a wide variety of viral, bacterial and fungal pathogens.Mix at a rate of 1.3 ounces per gallon of water. One gallon of solution treats 135 sq.ft.; 10-lb container makes 123 gallons of solution. Solution remains stable for seven days. EPA-registered (registration pending in California).