Learn More
MSC Vital Oxide Disinfectant, Liquid, Multi-surface, No-rinse, 5 gal
Catalog No. 502818022
Encompass
Vital Oxide is a versatile cleaner, disinfectant, mold controller, and odor eliminator. This hospital-grade disinfectant cleaner features a unique form of stabilized chlorine dioxide, effective at killing bacteria, viruses, and mold. It is non-corrosive, mild, and ready to use without mixing, suitable for a wide range of applications.
  • Kills 99.999% of bacteria on hard nonporous surfaces in 60 seconds
  • EPA registered (82972-1) and NSF listed (141392) for no-rinse use on food contact surfaces
  • Combats mold and mildew growth on hard surfaces and fabric
  • Non-corrosive and non-abrasive, safe for water-safe surfaces
  • Made without bleach and fragrance, with a low Category IV toxicity rating
Catalog No. 50-281-8022 Supplier MSC Supplier No. 14878078
Vital Oxide is a versatile cleaner, disinfectant, mold controller, and odor eliminator. This hospital-grade disinfectant cleaner features a unique form of stabilized chlorine dioxide, effective at killing bacteria, viruses, and mold. It is non-corrosive, mild, and ready to use without mixing, suitable for a wide range of applications.
  • Kills 99.999% of bacteria on hard nonporous surfaces in 60 seconds
  • EPA registered (82972-1) and NSF listed (141392) for no-rinse use on food contact surfaces
  • Combats mold and mildew growth on hard surfaces and fabric
  • Non-corrosive and non-abrasive, safe for water-safe surfaces
  • Made without bleach and fragrance, with a low Category IV toxicity rating