Learn More
MSC Contec Sporicidin Disinfectant Solution, Fresh Scent, EPA-Registered, 1.0 gal
Catalog No. 502819622
Encompass
Sporicidin Disinfectant Solution is a trusted cleaner and disinfectant for professional use, effective against disease-causing microbes and approved for use against SARS-CoV-2. It utilizes a powerful active ingredient system, including 1.56% Phenol, to eliminate pathogens and prevent odor-causing bacteria for up to six months. With an EPA Category IV rating, it offers very low toxicity for safe use in various environments, suitable for mold remediation and trauma scene cleanup.
  • EPA-registered bactericide, fungicide, virucide, and tuberculocide
  • Effective against a broad range of pathogens, including enveloped and non-enveloped viruses
  • Contains 1.56% Phenol (CAS 108-95-2)
  • Formulated with an EPA Category IV rating for very low acute toxicity
  • Disinfects, cleans, and deodorizes in one step
  • Miscible in water
Catalog No. 50-281-9622 Supplier MSC Supplier No. 06103493
Sporicidin Disinfectant Solution is a trusted cleaner and disinfectant for professional use, effective against disease-causing microbes and approved for use against SARS-CoV-2. It utilizes a powerful active ingredient system, including 1.56% Phenol, to eliminate pathogens and prevent odor-causing bacteria for up to six months. With an EPA Category IV rating, it offers very low toxicity for safe use in various environments, suitable for mold remediation and trauma scene cleanup.
  • EPA-registered bactericide, fungicide, virucide, and tuberculocide
  • Effective against a broad range of pathogens, including enveloped and non-enveloped viruses
  • Contains 1.56% Phenol (CAS 108-95-2)
  • Formulated with an EPA Category IV rating for very low acute toxicity
  • Disinfects, cleans, and deodorizes in one step
  • Miscible in water