Learn More
MSC Vital Oxide All-Purpose Cleaner, Disinfectant, Multi-Surface, NSF Compliant, 55 gal
Catalog No. 502819889
Encompass
Vital Oxide is a versatile, ready-to-use all-purpose cleaner and disinfectant, supplied in a 55 gal drum. It effectively eliminates allergens, odors, mold, and mildew, and is suitable for various applications in office and workplace settings.
  • EPA Registered as a Hospital Disinfectant
  • NSF Registered as a No-rinse required Food Surface Sanitizer
  • Proven to kill a wide range of viruses and bacteria
  • Contains no volatile organic compounds (VOCs)
  • No personal protective equipment (PPE) required for general cleaning
  • Sanitizes color-safe fabrics and carpets without damage
  • Compatible with electrostatic sprayers for efficient disinfection
  • Non-acidic and non-flammable formulation
Catalog No. 50-281-9889 Supplier MSC Supplier No. 17007576
Vital Oxide is a versatile, ready-to-use all-purpose cleaner and disinfectant, supplied in a 55 gal drum. It effectively eliminates allergens, odors, mold, and mildew, and is suitable for various applications in office and workplace settings.
  • EPA Registered as a Hospital Disinfectant
  • NSF Registered as a No-rinse required Food Surface Sanitizer
  • Proven to kill a wide range of viruses and bacteria
  • Contains no volatile organic compounds (VOCs)
  • No personal protective equipment (PPE) required for general cleaning
  • Sanitizes color-safe fabrics and carpets without damage
  • Compatible with electrostatic sprayers for efficient disinfection
  • Non-acidic and non-flammable formulation