Learn More
MSC ZEP All-Purpose Cleaner, Liquid, Disinfectant, 2 L, 4/Pk
Catalog No. 502818031
Encompass
ZEP DZ-7 is an all-purpose liquid cleaner and disinfectant designed to kill microbes on precleaned environmental surfaces. It has a pleasant scent and is suitable for various cleaning applications.
  • All-purpose cleaner and degreaser
  • Liquid form
  • Pleasant scent
  • Kills microbes on precleaned environmental surfaces
  • Disinfectant properties
  • Has a pH of 6–7
  • Water soluble
Catalog No. 50-281-8031 Supplier MSC Supplier No. 17642364
ZEP DZ-7 is an all-purpose liquid cleaner and disinfectant designed to kill microbes on precleaned environmental surfaces. It has a pleasant scent and is suitable for various cleaning applications.
  • All-purpose cleaner and degreaser
  • Liquid form
  • Pleasant scent
  • Kills microbes on precleaned environmental surfaces
  • Disinfectant properties
  • Has a pH of 6–7
  • Water soluble