Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC SC Johnson Professional Quaternary Disinfectant Cleaner, Fragrance Free, 32 oz

Catalog No. 502867203
Encompass

Quaternary Disinfectant Cleaner is an effective one-step virucide disinfectant cleaner for use on non-porous surfaces. It is effective against a broad spectrum of bacteria, is virucidal, fungicidal, and inhibits the growth of mold and mildew when used as directed. It also sanitizes soft surfaces as a spot treatment in 5 minutes.

  • One-step cleaner and disinfectant
  • Effective against a broad spectrum of bacteria, virucidal, fungicidal, and inhibits mold and mildew growth
  • Sanitizes soft surfaces as a spot treatment in 5 minutes
  • Kills SARS-CoV-2 virus (cause of COVID-19), Influenza A Virus, and Norovirus on hard, non-porous surfaces
  • Listed on the EPA's List Q for disinfectants against Monkey Pox
  • Fragrance free
  • PH 13
  • Density 1.005 g/cm³

Catalog No. 50-286-7203 Supplier MSC Supplier No. 17385535
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quaternary Disinfectant Cleaner is an effective one-step virucide disinfectant cleaner for use on non-porous surfaces. It is effective against a broad spectrum of bacteria, is virucidal, fungicidal, and inhibits the growth of mold and mildew when used as directed. It also sanitizes soft surfaces as a spot treatment in 5 minutes.

  • One-step cleaner and disinfectant
  • Effective against a broad spectrum of bacteria, virucidal, fungicidal, and inhibits mold and mildew growth
  • Sanitizes soft surfaces as a spot treatment in 5 minutes
  • Kills SARS-CoV-2 virus (cause of COVID-19), Influenza A Virus, and Norovirus on hard, non-porous surfaces
  • Listed on the EPA's List Q for disinfectants against Monkey Pox
  • Fragrance free
  • PH 13
  • Density 1.005 g/cm³

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.