Learn More
MSC Zep Disinfectant Cleaner, Odorless, Clear, 2-8 oz/gal, 1 gal.
Catalog No. 502815578
Encompass
Zep PDC Plain Disinfectant Cleaner II is formulated for effective disinfection of inanimate, hard, non-porous surfaces. It is suitable for use across various environments, from bathrooms and kitchens to healthcare facilities, ensuring comprehensive cleaning and pathogen control.
  • Odorless and clear, colorless formulation
  • Dilutable from 2 to 8 oz. per gallon for disinfection
  • Effective against a broad spectrum of viruses, including Canine Parvovirus, HIV-1, HBV, HCV, SARS, and Norwalk Virus
  • Targets common bacteria like E.Coli, MRSA, Pseudomonas aeruginosa, and Staphylococcus aureus
  • Prevents the growth of mold, mildew, and athlete's foot fungus
  • EPA registered for verified efficacy
Catalog No. 50-281-5578 Supplier MSC Supplier No. 17642166
Zep PDC Plain Disinfectant Cleaner II is formulated for effective disinfection of inanimate, hard, non-porous surfaces. It is suitable for use across various environments, from bathrooms and kitchens to healthcare facilities, ensuring comprehensive cleaning and pathogen control.
  • Odorless and clear, colorless formulation
  • Dilutable from 2 to 8 oz. per gallon for disinfection
  • Effective against a broad spectrum of viruses, including Canine Parvovirus, HIV-1, HBV, HCV, SARS, and Norwalk Virus
  • Targets common bacteria like E.Coli, MRSA, Pseudomonas aeruginosa, and Staphylococcus aureus
  • Prevents the growth of mold, mildew, and athlete's foot fungus
  • EPA registered for verified efficacy