Learn More
Bellco Glass, Inc. Disinfectant Cylinder,8x10mm
SDP
Catalog No. 501945423
Encompass_Preferred
For use in the disinfectant testing procedure as described in aqac manual. Cylinder ends are lightly glazed. Hard surface carrier test for efficacy of testing of disinfectants. MADE IN THE USA. Sizes: 8x10mm
Catalog No. 50-194-5423 Supplier Bellco Glass, Inc. Supplier No. 209108010
For use in the disinfectant testing procedure as described in aqac manual. Cylinder ends are lightly glazed. Hard surface carrier test for efficacy of testing of disinfectants. MADE IN THE USA. Sizes: 8x10mm