Learn More
MSC Pro-Series Wipers Disinfecting Wipes, Pre-Moistened, Refill Roll, 800 Sheets, White
Catalog No. 502838991
Encompass
Pro-Series Wipers Disinfecting Wipes are pre-moistened refill rolls designed for surface disinfecting in various environments. These white, lemon-scented wipes are alcohol-free and low lint, suitable for hospitality, healthcare, and office settings. Each pouch contains 800 sheets, providing a convenient solution for maintaining hygiene.
  • Pre-moistened flat sheets for surface disinfecting
  • Bucket refill packaging
  • Sheet dimensions: 6 inches long by 7 inches wide
  • Suitable for hospitality, healthcare, and office environments
  • White, lemon-scented wipes
  • Alcohol-free and low lint
  • 400 feet roll length, 1 ply
  • 800 sheets per pouch, 4 pouches per case
Catalog No. 50-283-8991 Supplier MSC Supplier No. 51728624
Pro-Series Wipers Disinfecting Wipes are pre-moistened refill rolls designed for surface disinfecting in various environments. These white, lemon-scented wipes are alcohol-free and low lint, suitable for hospitality, healthcare, and office settings. Each pouch contains 800 sheets, providing a convenient solution for maintaining hygiene.
  • Pre-moistened flat sheets for surface disinfecting
  • Bucket refill packaging
  • Sheet dimensions: 6 inches long by 7 inches wide
  • Suitable for hospitality, healthcare, and office environments
  • White, lemon-scented wipes
  • Alcohol-free and low lint
  • 400 feet roll length, 1 ply
  • 800 sheets per pouch, 4 pouches per case