Learn More
Med Vet International Cavicide Disinfectant Spray, 24 fl. oz.
Supplier:  Med Vet International CV1024
Encompass_Preferred
Cavicide is an effective, ready to use, multi-purpose, broad spectrum solution for use on the surfaces of inanimate objects. EPA reviews and registers antimicrobial pesticides, which include disinfectants for use on pathogens like SARS-CoV-2, the novel human coronavirus that causes COVID-19. EPA has been working to ensure that families, communities, businesses, hospitals, and others are aware of and have access to effective surface disinfectant products to use against SARS-CoV-2. EPA's List N contains disinfectants for Use Against SARS-CoV-2 (COVID-19). EPA expects all products on List N to be effective against SARS-CoV-2 (COVID-19) when used according to label directions. List N only includes EPA-registered surface disinfectants. This product has been tested for use against Covid-19 and is on List N. Active Ingredient: Quaternary ammonium; Isopropanol (Isopropyl alcohol) Contact Time in minutes: 2
Catalog No. 50-209-1784