Learn More
MSC CloroxPro Disinfectant & Sanitizer, Aerosol, 14 fl oz, Citrus
Catalog No. 502839978
Encompass
This aerosol disinfectant and sanitizer offers versatile use, effectively disinfecting hard surfaces, sanitizing soft surfaces, and eliminating odor. No pre-cleaning is required.
  • Disinfects hard surfaces
  • Sanitizes soft surfaces and air
  • No pre-cleaning required
  • Kills 24 germs on hard surfaces
  • Sanitizes soft surfaces in 30 seconds
  • Eliminates odor-causing bacteria
  • Reduces common allergens (e.g., dander)
  • Effective against SARS-CoV-2 (COVID-19 virus) per EPA policy
  • Accomplishes more with fewer cleaning products
  • Provides versatile aerosol application
  • Promotes healthier facilities
Catalog No. 50-283-9978 Supplier MSC Supplier No. 91482448
This aerosol disinfectant and sanitizer offers versatile use, effectively disinfecting hard surfaces, sanitizing soft surfaces, and eliminating odor. No pre-cleaning is required.
  • Disinfects hard surfaces
  • Sanitizes soft surfaces and air
  • No pre-cleaning required
  • Kills 24 germs on hard surfaces
  • Sanitizes soft surfaces in 30 seconds
  • Eliminates odor-causing bacteria
  • Reduces common allergens (e.g., dander)
  • Effective against SARS-CoV-2 (COVID-19 virus) per EPA policy
  • Accomplishes more with fewer cleaning products
  • Provides versatile aerosol application
  • Promotes healthier facilities