Learn More
Med Vet International Metrex Cavicide Wipes, 160 Wipes
Supplier:  Med Vet International CV1100
Encompass_Preferred
Presaturated with Cavicide, the cloth wipes are proven to kill both TB and Hepatitis B. Durable, non-woven, non-abrasive towels that dont bunch up during use. EPA reviews and registers antimicrobial pesticides, which include disinfectants for use on pathogens like SARS-CoV-2, the novel human coronavirus that causes COVID-19. EPA has been working to ensure that families, communities, businesses, hospitals, and others are aware of and have access to effective surface disinfectant products to use against SARS-CoV-2. EPA's List N contains disinfectants for Use Against SARS-CoV-2 (COVID-19). EPA expects all products on List N to be effective against SARS-CoV-2 (COVID-19) when used according to label directions. List N only includes EPA-registered surface disinfectants. This product has been tested for use against Covid-19 and is on List N. Active Ingredient: Quaternary ammonium; Isopropanol (Isopropyl alcohol) Contact Time in minutes: 3
Catalog No. 50-255-9603
 Restricted Item