Learn More
Med Vet International Cavicide Wipes, 160 Wipes/Canister, 12 Canisters/Case
Supplier:  Med Vet International CV1100CS
Encompass_Preferred
The power of Cavicide in a wipe! Presaturated with Cavicide, the cloth wipes are proven to kill both TB and Hepatitis B. Durable, non-woven, non-abrasive towels that dont bunch up during use. Does the job of three products- cleaner, disinfectant and decontaminant. 160 per tub. EPA reviews and registers antimicrobial pesticides, which include disinfectants for use on pathogens like SARS-CoV-2, the novel human coronavirus that causes COVID-19. EPA has been working to ensure that families, communities, businesses, hospitals, and others are aware of and have access to effective surface. This product has been tested for use against Covid-19 and is on List N. Active Ingredient: Quaternary ammonium; Isopropanol (Isopropyl alcohol) Contact Time in minutes: 3
Catalog No. 50-255-9602