Learn More
Grainger DISINFECTING WIPE,BUCKET,110CT
Supplier:  Grainger 4XKR7
Encompass_Preferred
Clorox Germicidal Disinfecting Wipes: Bucket, 110 ct Container Size, Ready to Use, 2 PKDisinfecting wipes are presaturated with a disinfectant that helps stop the spread of bacteria, fungi, and viruses. Use wipes after cleaning surfaces first to reduce the risk of spreading infection. They offer a convenient alternative to using multiple items, such as rags, buckets, and spray bottles. Disinfecting wipes lower the risk of cross-contamination on surfaces in restaurants, schools, gyms, and healthcare facilities.
Catalog No. NC2166421