Learn More
MSC Clorox Disinfecting Wipes, Bleach-Free, Fresh Scent, 450 Sheets
Catalog No. 502870560
Encompass
Clorox Disinfecting Wipes are EPA-registered wipes formulated to kill 99.9% of viruses and bacteria, including SARS-CoV-2, on hard, nonporous surfaces. They effectively remove allergens, clean messes, and provide 24-hour odor prevention.
  • EPA-registered formula ensures broad-spectrum efficacy
  • Kills 99.9% of bacteria in 10 seconds and most viruses in 15 seconds
  • Proven effective against SARS-CoV-2 on hard, nonporous surfaces
  • Removes common allergens and prevents odors for 24 hours
  • Bleach-free solution effectively breaks down grease, soap scum, and grime
  • Pre-moistened wipes provide convenient, single-swipe cleaning
Catalog No. 50-287-0560 Supplier MSC Supplier No. 91498808
Clorox Disinfecting Wipes are EPA-registered wipes formulated to kill 99.9% of viruses and bacteria, including SARS-CoV-2, on hard, nonporous surfaces. They effectively remove allergens, clean messes, and provide 24-hour odor prevention.
  • EPA-registered formula ensures broad-spectrum efficacy
  • Kills 99.9% of bacteria in 10 seconds and most viruses in 15 seconds
  • Proven effective against SARS-CoV-2 on hard, nonporous surfaces
  • Removes common allergens and prevents odors for 24 hours
  • Bleach-free solution effectively breaks down grease, soap scum, and grime
  • Pre-moistened wipes provide convenient, single-swipe cleaning