Learn More
MSC Diversey Disinfectant Cleaner, Ready-to-Use, Hospital-Grade, Lemon Scent, 32 oz, 12/Carton
Catalog No. 502870537
Encompass
Virex TB is a ready-to-use, quaternary-based hospital-grade disinfectant cleaner designed for effective cleaning and deodorizing of hard non-porous surfaces.
  • Disinfects in three minutes
  • Powerful bactericide, tuberculocide, virucide, and fungicide
  • EPA List N-approved to kill the COVID-19 virus
  • Cleans and deodorizes
  • Suitable for hard non-porous surfaces
Catalog No. 50-287-0537 Supplier MSC Supplier No. 91482562
Virex TB is a ready-to-use, quaternary-based hospital-grade disinfectant cleaner designed for effective cleaning and deodorizing of hard non-porous surfaces.
  • Disinfects in three minutes
  • Powerful bactericide, tuberculocide, virucide, and fungicide
  • EPA List N-approved to kill the COVID-19 virus
  • Cleans and deodorizes
  • Suitable for hard non-porous surfaces