Learn More
Medchemexpress LLC HY-P9985 5mg , Disitamab vedotin CAS:2136633-23-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99855MG
Encompass_Preferred
HY-P9985 5mg Disitamab vedotin CAS: 2136633-23-1 Disitamab vedotin (RC48) is an antibody-drug conjugate (ADC) comprising a monoclonal antibody against human epidermal growth factor receptor 2 (HER2) conjugated via a cleavable linker to the cytotoxic agent Monomethyl auristatin E (MMAE). Disitamab vedotin enhances antitumor immunity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1224