Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 1-(Dimethylphenylazo)-2-naphthol-3,6-disulfonate, TCI America™
SDP

Catalog No. P059125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P059125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P059125G Supplier TCI America Supplier No. P059125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00036432
Synonym 1-(Dimethylphenylazo)-2-naphthol-3,6-disulfonic Acid Disodium Salt

Specifications

Color Red
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Disodium 1-(Dimethylphenylazo)-2-naphthol-3,6-disulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.