Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 2,6-Naphthalenedisulfonate 90.0+%, TCI America™
SDP

Catalog No. N001325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N001325G 25 g
N0013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N001325G Supplier TCI America Supplier No. N001325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1655-45-4
Molecular Formula C10H6Na2O6S2
Molecular Weight (g/mol) 332.252
MDL Number MFCD00004092
InChI Key WZZLWPIYWZEJOX-UHFFFAOYSA-L
Synonym 2,6-naphthalenedisulfonic acid disodium salt, sodium naphthalene-2,6-disulfonate, disodium 2,6-naphthalenedisulfonate, disodium naphthalene-2,6-disulphonate, 2,6-naphthalenedisulfonic acid, disodium salt, ebert-merz beta-acid disodium salt, disodium naphthalene-2,6-disulfonate, dipotassium 2,6-naphthalenedisulfonate, acmc-209dsq, ksc910s4f
PubChem CID 3014027
IUPAC Name disodium;naphthalene-2,6-disulfonate
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)[O-])C=C1S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 332.25
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 2,6-Naphthalenedisulfonate
EINECSNumber (4)-0468
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.