Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 2,7-Naphthalenedisulfonate 98.0+%, TCI America™
SDP

Catalog No. N001425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N001425G 25 g
N0014500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N001425G Supplier TCI America Supplier No. N001425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1655-35-2
Molecular Formula C10H6Na2O6S2
Molecular Weight (g/mol) 332.252
MDL Number MFCD00066365
InChI Key XOIWXJSPLXGSLZ-UHFFFAOYSA-L
Synonym 2,7-Naphthalenedisulfonic Acid Disodium Salt, Ebert-Merz alpha-Acid Disodium Salt
PubChem CID 74249
IUPAC Name disodium;naphthalene-2,7-disulfonate
SMILES C1=CC(=CC2=C1C=CC(=C2)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 332.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 2,7-Naphthalenedisulfonate
EINECSNumber (4)-0468
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.