Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 2-Oxoglutarate 98.0+%, TCI America™
SDP

Catalog No. K000725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K000725G 25 g
K00075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K000725G Supplier TCI America Supplier No. K000725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 305-72-6
Molecular Formula C5H4Na2O5
Molecular Weight (g/mol) 190.062
MDL Number MFCD00043347
InChI Key YBGBJYVHJTVUSL-UHFFFAOYSA-L
Synonym disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate
PubChem CID 31040
IUPAC Name disodium;2-oxopentanedioate
SMILES C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 190.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 2-Oxoglutarate
RTECSNumber SA0454700
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.