Learn More
Selleck Chemical LLC Disodium
SDP
Catalog No. 5000003866
Encompass_Preferred
Disodium (R)-2-Hydroxyglutarate (D- -Hydroxyglutaric acid disodium) is a competitive inhibitor of -ketoglutarate-dependent dioxygenases with Ki of 10 87 1 85 mM
Catalog No. 50-000-03866 Supplier Selleck Chemical LLC Supplier No. S7873100MG
Disodium (R)-2-Hydroxyglutarate (D- -Hydroxyglutaric acid disodium) is a competitive inhibitor of -ketoglutarate-dependent dioxygenases with Ki of 10 87 1 85 mM