Learn More
Medchemexpress LLC Disodium 5'-inosinate | 4691-65-0 | 99.7% | 1 ML
SDP
Catalog No. 5000409967
Encompass_Preferred
Disodium 5'-inosinate is a nucleotide-based umami agent that binds to umami receptors and stimulates taste nerves, allowing the brain to perceive umami. It can also produce a transient behavioral excitement in mice.
  • Binds to umami receptors and stimulates taste nerves.
  • Produces transient behavioral excitement in mice.
  • Effective in stimulating oral responses in rat solitary-tract nucleus neurons, involved in encoding umami taste.
Catalog No. 50-004-09967 Supplier Medchemexpress LLC Supplier No. HYD088710MM/1ML
Disodium 5'-inosinate is a nucleotide-based umami agent that binds to umami receptors and stimulates taste nerves, allowing the brain to perceive umami. It can also produce a transient behavioral excitement in mice.
  • Binds to umami receptors and stimulates taste nerves.
  • Produces transient behavioral excitement in mice.
  • Effective in stimulating oral responses in rat solitary-tract nucleus neurons, involved in encoding umami taste.