Learn More
Medchemexpress LLC Disodium 5'-inosinate | 4691-65-0 | 99.7% | 25 G
SDP
Catalog No. 5000414464
Encompass_Preferred
Disodium 5'-inosinate is a nucleotide-based umami agent. It can bind to umami receptors and stimulate taste nerves, allowing the brain to perceive umami. In addition, it produces a transient behavioral excitement in mice.
  • Responds to oral stimulation in rat solitary-tract nucleus neurons, involved in encoding umami taste.
  • Produces transient behavioral excitement in mice.
  • In mice (500 mg/kg, intravenous injection), it led to increased movement, jumping, squeaking, enhanced preening, and increased reflex response, with sedation after 60 minutes.
Catalog No. 50-004-14464 Supplier Medchemexpress LLC Supplier No. 25GHYD0887
Add to Cart
Add to Cart
Disodium 5'-inosinate is a nucleotide-based umami agent. It can bind to umami receptors and stimulate taste nerves, allowing the brain to perceive umami. In addition, it produces a transient behavioral excitement in mice.
  • Responds to oral stimulation in rat solitary-tract nucleus neurons, involved in encoding umami taste.
  • Produces transient behavioral excitement in mice.
  • In mice (500 mg/kg, intravenous injection), it led to increased movement, jumping, squeaking, enhanced preening, and increased reflex response, with sedation after 60 minutes.