Learn More
Medchemexpress LLC Disodium 5'-inosinate (IMP disodium salt) | 4691-65-0 | 99.74% | 100 G
SDP
Catalog No. 5000410172
Encompass_Preferred
Disodium 5'-inosinate (IMP disodium salt) is a nucleotide-based umami agent. Disodium 5'-inosinate can bind to umami receptors and stimulate taste nerves, allowing the brain to perceive umami. In addition, Disodium 5'-inosinate produces a transient behavioral excitement in mice.
  • Responds to oral stimulation in rat solitary-tract nucleus neurons and is involved in encoding umami taste.
  • Produced a behavioral excitement in mice showing an increased movement, jumping and squeaking in response to tactile stimuli and enhanced preening and increased reflex response.
Catalog No. 50-004-10172 Supplier Medchemexpress LLC Supplier No. HYD0887100G
Add to Cart
Add to Cart
Disodium 5'-inosinate (IMP disodium salt) is a nucleotide-based umami agent. Disodium 5'-inosinate can bind to umami receptors and stimulate taste nerves, allowing the brain to perceive umami. In addition, Disodium 5'-inosinate produces a transient behavioral excitement in mice.
  • Responds to oral stimulation in rat solitary-tract nucleus neurons and is involved in encoding umami taste.
  • Produced a behavioral excitement in mice showing an increased movement, jumping and squeaking in response to tactile stimuli and enhanced preening and increased reflex response.