Learn More
Medchemexpress LLC Disodium 6,6′-oxybis(2-naphthalenesulfonate) | 61551-82-4 | N/A | >90.0% | 474.4 | C20H12Na2O7S2 | 5 MG
SDP
Catalog No. 5000284556
Encompass_Preferred
Disodium 6,6′-oxybis(2-naphthalenesulfonate) is a disodium salt supplied for research and analytical use. It functions as a surfactant, emulsifier, and dispersant, and is reported as an impurity of Sunset Yellow FCF. The compound is used as an intermediate or analytical standard in polymer, pharmaceutical, and agrochemical synthesis.
Catalog No. 50-002-84556 Supplier Medchemexpress LLC Supplier No. HYW5858225MG
Disodium 6,6′-oxybis(2-naphthalenesulfonate) is a disodium salt supplied for research and analytical use. It functions as a surfactant, emulsifier, and dispersant, and is reported as an impurity of Sunset Yellow FCF. The compound is used as an intermediate or analytical standard in polymer, pharmaceutical, and agrochemical synthesis.