Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium Adipate 98.0+%, TCI America™
SDP

Catalog No. A0592500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0592500G 500 g
A059225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0592500G Supplier TCI America Supplier No. A0592500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7486-38-6
Molecular Formula C6H8Na2O4
Molecular Weight (g/mol) 190.106
MDL Number MFCD00070498
InChI Key KYKFCSHPTAVNJD-UHFFFAOYSA-L
Synonym Adipic Acid Disodium Salt
PubChem CID 24073
IUPAC Name disodium;hexanedioate
SMILES C(CCC(=O)[O-])CC(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 190.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Adipate
EINECSNumber (2)-0859
RTECSNumber AV1770000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.