Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disodium cantharidin | 1465-77-6 | 95.0% | 258.18 g·mol⁻1 | C10H12Na2O5 | 5 MG
SDP

Catalog No. 5000222468
Encompass_Preferred

Disodium cantharidin is the disodium salt of cantharidin and functions as a selective inhibitor of protein phosphatases PP2A and PP1. Supplied as a solid research reagent (5 mg), it is used in biochemical and cell-based studies to probe phosphatase activity, cell signaling, and apoptosis. For research use only.

  • Selective PP2A and PP1 inhibitor, useful for signaling studies.
  • Reported activity: PP2A IC50 ≈ 50 nM, PP1 IC50 ≈ 600 nM.
  • High purity solid, typically 95% by vendor specification.
  • Small package sizes suitable for screening and method development.
  • Sodium salt form with formula C10H12Na2O5 and molecular weight 258.18 g·mol⁻1.

Catalog No. 50-002-22468 Supplier Medchemexpress LLC Supplier No. HY137135A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disodium cantharidin is the disodium salt of cantharidin and functions as a selective inhibitor of protein phosphatases PP2A and PP1. Supplied as a solid research reagent (5 mg), it is used in biochemical and cell-based studies to probe phosphatase activity, cell signaling, and apoptosis. For research use only.

  • Selective PP2A and PP1 inhibitor, useful for signaling studies.
  • Reported activity: PP2A IC50 ≈ 50 nM, PP1 IC50 ≈ 600 nM.
  • High purity solid, typically 95% by vendor specification.
  • Small package sizes suitable for screening and method development.
  • Sodium salt form with formula C10H12Na2O5 and molecular weight 258.18 g·mol⁻1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.