Learn More
Medchemexpress LLC Disodium creatine phosphate | 10 G
SDP
Catalog No. 5000409873
Encompass_Preferred
Phosphocreatine (creatine phosphate) is an organic compound found in vertebrate skeletal muscles. It enhances antioxidant activity and activates the TAK1 pathway to protect the heart. Phosphocreatine normalizes mitochondrial function and reduces oxidative stress via the Akt-mediated Nrf2/HO-1 pathway. It also provides renal protection by suppressing Apoptosis and ROS (Reactive Oxygen Species) generation through the ERK-mediated Nrf-2/HO-1 pathway.
  • Enhances antioxidant activity
  • Activates the TAK1 pathway to protect the heart
  • Normalizes mitochondrial function
  • Reduces oxidative stress via the Akt-mediated Nrf2/HO-1 pathway
  • Provides renal protection by suppressing Apoptosis and ROS generation through the ERK-mediated Nrf-2/HO-1 pathway
Catalog No. 50-004-09873 Supplier Medchemexpress LLC Supplier No. HYD088510G
Add to Cart
Add to Cart
Phosphocreatine (creatine phosphate) is an organic compound found in vertebrate skeletal muscles. It enhances antioxidant activity and activates the TAK1 pathway to protect the heart. Phosphocreatine normalizes mitochondrial function and reduces oxidative stress via the Akt-mediated Nrf2/HO-1 pathway. It also provides renal protection by suppressing Apoptosis and ROS (Reactive Oxygen Species) generation through the ERK-mediated Nrf-2/HO-1 pathway.
  • Enhances antioxidant activity
  • Activates the TAK1 pathway to protect the heart
  • Normalizes mitochondrial function
  • Reduces oxidative stress via the Akt-mediated Nrf2/HO-1 pathway
  • Provides renal protection by suppressing Apoptosis and ROS generation through the ERK-mediated Nrf-2/HO-1 pathway