Learn More
Selleck Chemical LLC Disodium Cromoglycate S1911-1g
SDP
Catalog No. 503110582
Encompass_Preferred
Disodium Cromoglycate (Gynazole FPL 670 (Cromolyn) Disodium Cromolyn sodium) is an antiallergic drug with IC50 of 39 g/mL
Catalog No. 50-311-0582 Supplier Selleck Chemical LLC Supplier No. S19111G
Disodium Cromoglycate (Gynazole FPL 670 (Cromolyn) Disodium Cromolyn sodium) is an antiallergic drug with IC50 of 39 g/mL