Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium Dimercaptomaleonitrile 90.0+%, TCI America™
SDP

Catalog No. D19301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19301G 1 g
D19305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19301G Supplier TCI America Supplier No. D19301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5466-54-6
Molecular Formula C4H2N2Na2OS2
Molecular Weight (g/mol) 204.173
MDL Number MFCD00060122
InChI Key DWYJCSXARKJSBC-FGSKAQBVSA-L
Synonym Dimercaptomaleonitrile Disodium Salt
PubChem CID 131675251
IUPAC Name disodium;(Z)-1,2-dicyanoethene-1,2-dithiolate;hydrate
SMILES C(#N)C(=C(C#N)[S-])[S-].O.[Na+].[Na+]

Specifications

Color Yellow
Quantity 1 g
UN Number 3439
Formula Weight 186.16
Percent Purity ≥90.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Dimercaptomaleonitrile
EINECSNumber (2)-2784
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.