Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylenediaminetetraacetic acid disodium salt dihydrate, 6381-92-6, 100g
Catalog No. NC0905064
Encompass
Empirical Formula (Hill Notation) C10H14N2Na2O8 · 2H2O, 99.0-101.0% (titration), Molecular Weight 372.24,mp 248 °C (dec.) (lit.), Synonym: Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2; Ethylenediaminetetraacetic acid disodium salt dehydrate is designated as Molecular Biology grade and is suitable for molecular biology applications. It has been analyzed for the presence of nucleases and proteases.
Catalog No. NC0905064 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E5134100G
Empirical Formula (Hill Notation) C10H14N2Na2O8 · 2H2O, 99.0-101.0% (titration), Molecular Weight 372.24,mp 248 °C (dec.) (lit.), Synonym: Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2; Ethylenediaminetetraacetic acid disodium salt dehydrate is designated as Molecular Biology grade and is suitable for molecular biology applications. It has been analyzed for the presence of nucleases and proteases.