Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium Glutarate, Spectrum™ Chemical
SDP

Catalog No. 18620566 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-566 100 g
18-620-565 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-566 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1540100GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: Glutaric Acid Disodium Salt, Disodium Glutarate

Chemical Identifiers

CAS 13521-83-0
Molecular Formula C5H6Na2O4
Molecular Weight (g/mol) 176.08
InChI Key ZUDYLZOBWIAUPC-UHFFFAOYSA-L
IUPAC Name disodium pentanedioate
SMILES [Na+].[Na+].[O-]C(=O)CCCC([O-])=O

Specifications

Quantity 100 g
Percent Purity 99%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Disodium Glutarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.