Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium hydrogen citrate sesquihydrate, 99%

Catalog No. AAA140010I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA140010I 5000 g
AAA1400130 250 g
AAA140010B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA140010I Supplier Thermo Scientific Chemicals Supplier No. A140010I
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 6132-05-4 | C12H18Na4O17 | 526.22 g/mol

Disodium hydrogen citrate sesquihydrate is a reagent employed in the investigation of mediatorless laccase cathodes as half-cell electrode under galvanostatic load by using a two-compartment set up. It is used in the extraction of organ/tissue sample.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disodium hydrogen citrate sesquihydrate is a reagent employed in the investigation of mediatorless laccase cathodes as half-cell electrode under galvanostatic load by using a two-compartment set up. It is used in the extraction of organ/tissue sample.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-4-6132
Molecular Formula C12H18Na4O17
Molecular Weight (g/mol) 526.22
MDL Number MFCD00150445
InChI Key HGPVLOQNBSHYEI-UHFFFAOYNA-J
Synonym unii-cz1032cekr, cz1032cekr, sodium citrate dibasic sesquihydrate, disodium citrate sesquihydrate, disodium hydrogen citrate 1.5 h2o, sodium hydrogen citrate sesquihydrate, citric acid, disodium salt, sesquihydrate, tetrasodium, 3-carboxy-3-hydroxypentanedioate, trihydrate, sodium citrate dibasic sesquihydrate, purum p.a t, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, disodium salt, hydrate 2:3
PubChem CID 117063816
IUPAC Name tetrasodium;2-(carboxymethyl)-2-hydroxybutanedioate;trihydrate
SMILES C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 149°C (decomposition)
Quantity 5000 g
Solubility Information Soluble in water.
Formula Weight 263.11 (236.09 Anhydrous)
Percent Purity 99%
Chemical Name or Material Disodium hydrogen citrate sesquihydrate
EINECSNumber 205-623-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.