Learn More
Medchemexpress LLC Disodium hydrogen phosphate anhydrous | 7558-79-4 | MFCD00003496 | 99.0% | 141.96 g/mol | Na2HPO4 | 1g
SDP
Catalog No. 5000269810
Encompass_Preferred
Disodium phosphate is commonly used as a food additive buffer and laboratory reagent Disodium phosphate has unique chemical properties that make it an important ingredient in the manufacture of fertilizers detergents and water treatment products Additionally it is used in various medical applications such as osmotic laxatives and electrolyte replacement solutions
Catalog No. 50-002-69810 Supplier Medchemexpress LLC Supplier No. HYY03081G
Disodium phosphate is commonly used as a food additive buffer and laboratory reagent Disodium phosphate has unique chemical properties that make it an important ingredient in the manufacture of fertilizers detergents and water treatment products Additionally it is used in various medical applications such as osmotic laxatives and electrolyte replacement solutions