Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium Iminodiacetate Hydrate 98.0+%, TCI America™
SDP

Catalog No. I000725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I000725G 25 g
I0007250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I000725G Supplier TCI America Supplier No. I000725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 207398-95-6
Molecular Formula C4H7NNa2O5
Molecular Weight (g/mol) 195.082
MDL Number MFCD00043340
InChI Key YSBGCHXLMBAKPV-UHFFFAOYSA-L
Synonym Iminodiacetic Acid Disodium Salt
PubChem CID 12520792
IUPAC Name disodium;2-(carboxylatomethylamino)acetate;hydrate
SMILES C(C(=O)[O-])NCC(=O)[O-].O.[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 177.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Iminodiacetate Hydrate
EINECSNumber (2)-1281
RTECSNumber AI2975500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.