Learn More
Medchemexpress LLC Disodium monofluorophosphate | 10163-15-2 | 10 G
SDP
Catalog No. 5000408883
Encompass_Preferred
Disodium monofluorophosphate is a biochemical reagent used as a biological material or organic compound for life science-related research. This product is for research use only.
  • Used as a biological material
  • Used as an organic compound for life science-related research
  • Solid appearance
  • White to off-white color
  • Soluble in water (≥ 100 mg/mL)
Catalog No. 50-004-08883 Supplier Medchemexpress LLC Supplier No. HYB054010G
Disodium monofluorophosphate is a biochemical reagent used as a biological material or organic compound for life science-related research. This product is for research use only.
  • Used as a biological material
  • Used as an organic compound for life science-related research
  • Solid appearance
  • White to off-white color
  • Soluble in water (≥ 100 mg/mL)