Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Disodium monofluorophosphate | 10163-15-2 | 100 G
SDP

Catalog No. 5000405853
Encompass_Preferred

Disodium monofluorophosphate is a biochemical reagent for life science-related research, suitable for use as a biological material or organic compound. It appears as a white to off-white solid.

  • Used as a biochemical reagent
  • Suitable for life science research
  • Solid appearance
  • White to off-white color
  • Soluble in water: ≥ 100 mg/mL (694.69 mM)
  • Store at 4°C, sealed and away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Catalog No. 50-004-05853 Supplier Medchemexpress LLC Supplier No. HYB0540100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disodium monofluorophosphate is a biochemical reagent for life science-related research, suitable for use as a biological material or organic compound. It appears as a white to off-white solid.

  • Used as a biochemical reagent
  • Suitable for life science research
  • Solid appearance
  • White to off-white color
  • Soluble in water: ≥ 100 mg/mL (694.69 mM)
  • Store at 4°C, sealed and away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.